Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01328428_s1
          See other VMN1R56 GE Assays ›
          Gene Symbol:
          Vmn1r56
          Gene Name:
          vomeronasal 1 receptor 56
          Gene Aliases:
          V1rd3, V3R3
          Chromosome Location:
          Chr.7: 5194916 - 5196747 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01328428_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vmn1r56
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_030740.1 NP_109665.1 1 1334 91
          GenBank mRNA AF324868.1 - 1 1334 91

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Vmn1r56

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P123209 NM_030740.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P123209 AF324868.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P123209 NM_030740.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P123209 AF324868.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P490605 NM_030740.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P490605 AF324868.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P490605 NM_030740.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P490605 AF324868.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952366 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24952376 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32983892 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32983880 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P32983893 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10559726 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559727 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559725 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559744 - -
          Affymetrix HT MG-430 PM Array Plate 1421736_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10559724 - -
          Affymetrix Mouse Gene 1.1 ST Array 10559743 - -
          Mouse Genome 430 2.0 Array 1421736_at - -
          Mouse Genome 430 2.0 Array 1421736_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 81015_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 81015_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700019994.mm.1 TC0700002189.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700019994.mm.1 NM_030740.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700019994.mm.1 NM_030740.1 Refseq
          Mouse Expression Set 430 1421736_PM_at - -
          Mouse Expression Set 430 1421736_at - -
          Mouse Genome 430A 2.0 Array 1421736_PM_at - -
          Mouse Genome 430A 2.0 Array 1421736_at - -
          Affymetrix HT Mouse Genome 430A Array 1421736_at - -
          Affymetrix HT Mouse Genome 430A Array 1421736_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 845264 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 630736 - -
          Illumina mouse-6 v1.0 expression beadchip 7040047 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000630736 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0007040047 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2435096 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1257296 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1257296 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2435096 - -
          mouse-6 v1.1 (Illumina) scl0081015.2_175-S - -
          mouse-6 v1.1 (Illumina) scl081015.1_95-S - -
          Illumina Sentrix mouse-6.v1 bead array 7040047 - -
          Illumina Sentrix mouse-6.v1 bead array 630736 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl081015.1_95-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0081015.2_175-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 630736 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 7040047 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1257296 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2435096 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2435096 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1257296 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline