Search
Search
| Gene Symbol: Epn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252454.1 | NP_001239383.1 | 2 - 3 | 500 | 75 | |
| NM_010147.4 | NP_034277.1 | 3 - 4 | 671 | 75 | ||
| GenBank mRNA | AK155033.1 | - | 3 - 4 | 666 | 75 | |
| BC046962.1 | - | 1 - 2 | 306 | 6391968 | 75 | |
| BC067206.1 | - | 2 - 3 | 442 | 6514327 | 75 | |
| BC099682.1 | - | 3 - 4 | 601 | 6529672 | 75 | |
Set Membership: |
|