Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200037049.mm.1 |
NM_026574.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037049.mm.1 |
NM_026574.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037049.mm.1 |
TC0200004433.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200037040.mm.1 |
NM_026574.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037040.mm.1 |
NM_026574.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037040.mm.1 |
TC0200004433.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200037045.mm.1 |
NM_026574.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037045.mm.1 |
NM_026574.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037045.mm.1 |
TC0200004433.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200037046.mm.1 |
NM_026574.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037046.mm.1 |
NM_026574.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037046.mm.1 |
TC0200004433.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200037048.mm.1 |
NM_026574.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037048.mm.1 |
NM_026574.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037048.mm.1 |
TC0200004433.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200037050.mm.1 |
NM_026574.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037050.mm.1 |
NM_026574.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037050.mm.1 |
TC0200004433.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200037051.mm.1 |
NM_026574.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037051.mm.1 |
NM_026574.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037051.mm.1 |
TC0200004433.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200037039.mm.1 |
NM_026574.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037039.mm.1 |
NM_026574.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037039.mm.1 |
TC0200004433.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200037041.mm.1 |
NM_026574.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037041.mm.1 |
NM_026574.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037041.mm.1 |
TC0200004433.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200037042.mm.1 |
NM_026574.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037042.mm.1 |
NM_026574.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037042.mm.1 |
TC0200004433.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200037044.mm.1 |
NM_026574.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037044.mm.1 |
NM_026574.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037044.mm.1 |
TC0200004433.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200037047.mm.1 |
NM_026574.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037047.mm.1 |
NM_026574.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037047.mm.1 |
TC0200004433.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200037043.mm.1 |
NM_026574.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037043.mm.1 |
NM_026574.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200037043.mm.1 |
TC0200004433.mm.1 |
MTA TC |