Search
Search

Ckap5| Gene Symbol: Ckap5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001165989.1 | NP_001159461.1 | 26 - 27 | 3451 | 62 | |
| NM_029437.2 | NP_083713.2 | 26 - 27 | 3451 | 62 | ||
| GenBank mRNA | AK122204.1 | - | 20 - 21 | 2454 | 62 | |
| BC089032.1 | - | 26 - 27 | 3465 | 30674362 | 62 | |
| BC096422.1 | - | 26 - 27 | 3432 | 30655413 | 62 | |
| BC138334.1 | - | 26 - 27 | 3437 | 8861354 | 62 | |
Set Membership: |
|