Search
Search

Apbb1ip| Gene Symbol: Apbb1ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019456.2 | NP_062329.2 | 2 - 3 | 266 | 83 | |
| GenBank mRNA | AK021331.1 | - | 3 - 4 | 145 | 83 | |
| AK040084.1 | - | 2 - 3 | 201 | 83 | ||
| AK041552.1 | - | 2 - 3 | 269 | 83 | ||
| AK042467.1 | - | 2 - 3 | 204 | 83 | ||
| AK079736.1 | - | 2 - 3 | 500 | 83 | ||
| AK162323.1 | - | 2 - 3 | 150 | 83 | ||
| BB638237.1 | - | 2 - 3 | 268 | 83 | ||
| BC023110.1 | - | 2 - 3 | 161 | 3707478 | 83 | |
Set Membership: |
|