Search
Search
| Gene Symbol: Oxt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011025.4 | NP_035155.1 | 2 - 3 | 368 | 79 | |
| GenBank mRNA | AF483508.1 | - | 2 - 3 | 327 | 79 | |
| AF483509.1 | - | 2 - 3 | 327 | 79 | ||
| AK003042.1 | - | 2 - 3 | 365 | 79 | ||
| AW455577.1 | - | 1 - 2 | 79 | |||
| BC117030.1 | - | 2 - 3 | 40126349 | 79 | ||
| BC117032.1 | - | 2 - 3 | 40126351 | 79 | ||
Set Membership: |
|