Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01329604_g1
          See other SPEF1 GE Assays ›
          Gene Symbol:
          Spef1
          Gene Name:
          sperm flagellar 1
          Gene Aliases:
          4931426K16Rik, Clamp
          Chromosome Location:
          Chr.2: 131170261 - 131174810 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01329604_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Spef1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027641.2 NP_081917.1 6 - 7 734 61
          GenBank mRNA AK016476.1 - 6 - 7 754 61
          AK046964.1 - 800 61
          AK047675.1 - 6 - 7 735 61
          AY860964.1 - 6 - 7 752 61
          BC006046.1 - 6 - 7 674 3590641 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200004650.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21633643 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21633642 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21633644 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21633641 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P299632 AK046964.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P299632 AK047675.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P299632 NM_027641.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P299632 AK046964.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P299632 AK047675.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P299632 NM_027641.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 190702 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 430752 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 132919 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 114765 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P115968 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P149145 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P118705 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P118620 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P299632 AK046964.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P299632 AK047675.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P299632 NM_027641.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P299632 AK046964.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P299632 AK047675.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P299632 NM_027641.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P149145 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P118705 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P115968 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P118620 - -
          Whole Mouse Genome G4122A A_52_P156765 BC006046.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P156765 AK046964.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P156765 AK047675.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P156765 AY860964.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P156765 NM_027641.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P118620 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P149145 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P118705 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P115968 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21633641 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21633643 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21633644 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21633642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21633655 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01723342 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01853795 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21633661 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21633636 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21633668 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21633643 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01443780 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21633635 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01360734 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21633659 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21633650 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21633674 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21633669 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21633662 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21633649 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21633637 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01723339 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21633671 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01853788 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21633681 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21633633 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21633677 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21633646 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21633670 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21633653 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01443774 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21633682 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21633647 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01443785 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21633631 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01360735 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21633665 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10487867 - -
          Affymetrix Mouse Gene 1.0 ST Array 10487866 - -
          Affymetrix Mouse Gene 1.0 ST Array 10487859 - -
          Affymetrix Mouse Gene 1.0 ST Array 10487865 - -
          Affymetrix Mouse Gene 1.0 ST Array 10487868 - -
          Affymetrix Mouse Gene 1.0 ST Array 10487864 - -
          Affymetrix Mouse Gene 1.0 ST Array 10487869 - -
          Affymetrix Mouse Gene 1.0 ST Array 10487860 - -
          Affymetrix Mouse Gene 1.0 ST Array 10487862 - -
          Affymetrix Mouse Gene 1.0 ST Array 10487861 - -
          Affymetrix HT MG-430 PM Array Plate 1424817_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1444397_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10487858 - -
          Mouse Genome 430 2.0 Array 1444397_PM_at - -
          Mouse Genome 430 2.0 Array 1424817_at - -
          Mouse Genome 430 2.0 Array 1444397_at - -
          Mouse Genome 430 2.0 Array 1424817_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70997_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70997_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0200039955.mm.1 NM_027641.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200039955.mm.1 NM_027641.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200039955.mm.1 TC0200004650.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200039953.mm.1 NM_027641.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200039953.mm.1 NM_027641.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200039953.mm.1 TC0200004650.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200039954.mm.1 NM_027641.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200039954.mm.1 NM_027641.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200039954.mm.1 TC0200004650.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200039956.mm.1 NM_027641.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200039956.mm.1 NM_027641.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200039956.mm.1 TC0200004650.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200039957.mm.1 NM_027641.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200039957.mm.1 NM_027641.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200039957.mm.1 TC0200004650.mm.1 MTA TC
          Mouse Expression Set 430 1424817_PM_at - -
          Mouse Expression Set 430 1424817_at - -
          Mouse Expression Set 430 1444397_PM_at - -
          Mouse Expression Set 430 1444397_at - -
          Murine Genome U74v2 Set 99012_PM_at - -
          Murine Genome U74v2 Set 99012_at - -
          Murine Genome U74v2 Set 93475_at - -
          Murine Genome U74v2 Set 93475_PM_at - -
          Mouse Genome 430A 2.0 Array 1424817_PM_at - -
          Mouse Genome 430A 2.0 Array 1424817_at - -
          Affymetrix HT Mouse Genome 430A Array 1424817_at - -
          Affymetrix HT Mouse Genome 430A Array 1424817_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1444397_at - -
          Affymetrix HT Mouse Genome 430B Array 1444397_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 743577 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1225629 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2701130 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2951596 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2951596 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline