Search
Search

1700037H04Rik| Gene Symbol: 1700037H04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026091.2 | NP_080367.1 | 3 - 4 | 330 | 68 | |
| GenBank mRNA | AK003406.1 | - | 3 - 4 | 331 | 68 | |
| AK006622.1 | - | 3 - 4 | 400 | 68 | ||
| AK178717.1 | - | 1 - 2 | 78 | 68 | ||
| BC006041.1 | - | 2 - 3 | 242 | 3497476 | 68 | |
Set Membership: |
|