Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01330423_g1
          See other LCN12 GE Assays ›
          Gene Symbol:
          Lcn12
          Gene Name:
          lipocalin 12
          Gene Aliases:
          9230102M18Rik
          Chromosome Location:
          Chr.2: 25490845 - 25494061 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01330423_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lcn12
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029958.1 NP_084234.1 4 - 5 510 89
          GenBank mRNA AK020307.1 - 3 - 4 416 89
          AY299493.1 - 4 - 5 510 89
          BC111100.1 - 4 - 5 503 40049540 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200003155.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lcn12

          Panther Classification:

          Molecular Function -

          transfer/carrier protein

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          biological_process
          retinoic acid binding
          transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1964509 AK020307.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1964509 AY299493.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1964509 NM_029958.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2135149 AY299493.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2135149 BC111100.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2135149 NM_029958.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 284857 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 883360 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P101469 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2135149 AY299493.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2135149 BC111100.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2135149 NM_029958.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1964509 AK020307.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1964509 AY299493.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1964509 NM_029958.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21072554 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21072538 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21072561 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21072543 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21072577 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21072570 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21072575 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21072563 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21072573 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21072552 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21072580 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21072539 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21072557 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21072544 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21072548 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21072542 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21072531 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21072532 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21072529 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21072545 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21072550 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21072582 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21072553 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21072536 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21072559 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21072533 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21072572 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21072547 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21072565 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21072534 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21072566 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21072556 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21072568 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21072540 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21072576 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P101469 - -
          Whole Mouse Genome G4122A A_51_P193640 AK020307.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P193640 AY299493.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P193640 BC111100.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P193640 NM_029958.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P101469 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01772359 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01772348 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01772363 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01772357 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01772367 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01772356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21072512 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21072580 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01959916 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21072524 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21072568 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21072515 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21072534 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21072504 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21072545 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21072542 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21072536 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21072552 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01723884 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21072528 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21072566 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21072559 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21072525 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21072517 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21072554 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01772376 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01772350 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21072563 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21072547 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21072521 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21072561 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01772354 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21072529 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21072570 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21072502 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21072533 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21072498 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01772357 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21072532 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02003006 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01412370 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01376692 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01772351 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01959919 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01772363 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02002715 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21072569 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02002713 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21072507 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21072582 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01723881 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01772367 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01723883 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21072576 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21072539 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01959924 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01959950 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10480746 - -
          Affymetrix Mouse Gene 1.0 ST Array 10480749 - -
          Affymetrix Mouse Gene 1.0 ST Array 10480748 - -
          Affymetrix Mouse Gene 1.0 ST Array 10480747 - -
          Affymetrix Mouse Gene 1.0 ST Array 10480750 - -
          Affymetrix Mouse Gene 1.0 ST Array 10480745 - -
          Affymetrix HT MG-430 PM Array Plate 1429935_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10480744 - -
          Mouse Genome 430 2.0 Array 1429935_PM_at - -
          Mouse Genome 430 2.0 Array 1429935_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 77701_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 77701_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0200026070.mm.1 NM_029958.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200026070.mm.1 NM_029958.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200026070.mm.1 TC0200003155.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200026071.mm.1 NM_029958.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200026071.mm.1 NM_029958.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200026071.mm.1 TC0200003155.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200026073.mm.1 NM_029958.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200026073.mm.1 NM_029958.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200026073.mm.1 TC0200003155.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200026074.mm.1 NM_029958.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200026074.mm.1 NM_029958.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200026074.mm.1 TC0200003155.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200026075.mm.1 NM_029958.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200026075.mm.1 NM_029958.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200026075.mm.1 TC0200003155.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200026072.mm.1 NM_029958.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200026072.mm.1 NM_029958.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200026072.mm.1 TC0200003155.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200026077.mm.1 NM_029958.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200026077.mm.1 NM_029958.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200026077.mm.1 TC0200003155.mm.1 MTA TC
          Mouse Expression Set 430 1429935_PM_at - -
          Mouse Expression Set 430 1429935_at - -
          Affymetrix HT Mouse Genome 430B Array 1429935_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1429935_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 311396 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2660461 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2660461 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline