Search
Search

Ppp6r1| Gene Symbol: Ppp6r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172894.2 | NP_766482.2 | 5 - 6 | 1094 | 67 | |
| GenBank mRNA | AK080891.1 | - | 5 - 6 | 1135 | 67 | |
| AK122450.1 | - | 5 - 6 | 1094 | 67 | ||
| BC053076.1 | - | 5 - 6 | 1094 | 5698212 | 67 | |
| BC141039.1 | - | 4 - 5 | 641 | 9055651 | 67 | |
Set Membership: |
|