Search
Search
| Gene Symbol: Ermn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029972.3 | NP_084248.2 | 1 - 2 | 279 | 69 | |
| GenBank mRNA | AK020733.1 | - | 1 - 2 | 391 | 69 | |
| AK078089.1 | - | 1 - 2 | 369 | 69 | ||
| AK136820.1 | - | 1 - 2 | 369 | 69 | ||
| AK137305.1 | - | 1 - 2 | 371 | 69 | ||
| AK220372.1 | - | 1 - 2 | 398 | 69 | ||
| BC089515.1 | - | 1 - 2 | 280 | 6589784 | 69 | |
| BC090670.1 | - | 1 - 2 | 311 | 30941185 | 69 | |
| DQ334270.1 | - | 1 - 2 | 257 | 69 | ||
Set Membership: |
|