Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01332105_m1
          See other NXT2 GE Assays ›
          Gene Symbol:
          Nxt2
          Gene Name:
          nuclear transport factor 2-like export factor 2
          Gene Aliases:
          6330587F24Rik, AI787442, P15-2
          Chromosome Location:
          Chr.X: 142226770 - 142239700 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01332105_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nxt2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001161430.2 NP_001154902.1 4 - 5 405 87
          NM_001290532.1 NP_001277461.1 4 - 5 490 87
          NM_001290533.1 NP_001277462.1 3 - 4 496 87
          NM_172782.4 NP_766370.2 3 - 4 442 87
          GenBank mRNA AK032102.1 - 3 - 4 413 87
          AK144349.1 - 3 - 4 426 87
          BC064727.1 - 3 - 4 417 87
          BC068166.1 - 3 - 4 395 30361776 87
          BC094570.1 - 3 - 4 359 30361291 87
          BC118503.1 - 3 - 4 370 40106658 87
          CA482101.1 - 4 - 5 441 87
          CJ148018.1 - 4 - 5 491 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00001402.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nxt2

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          biological_process
          protein transport
          mRNA transport
          protein binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1990041 BC068166.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1990041 BC094570.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1990041 AK144349.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1990041 BC064727.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1990041 NM_172782.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 568210 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 923907 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 345612 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 918616 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 543544 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 144187 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 127094 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P141416 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P157507 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1990041 BC068166.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1990041 BC094570.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1990041 AK144349.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1990041 BC064727.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1990041 NM_172782.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P517904 BC068166.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P517904 BC094570.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P517904 BC118503.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P517904 AK144349.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P517904 BC064727.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32707027 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32707036 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32707045 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32707053 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32707028 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32707050 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32707059 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32707037 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32707031 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32707051 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32707032 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32707062 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32707044 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32707048 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32707026 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32707055 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32707054 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32707061 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32707039 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32707041 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32707057 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32707030 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32707035 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32707064 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32707042 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32707058 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32707025 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32707046 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32707056 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P141416 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P157507 - -
          Whole Mouse Genome G4122A A_51_P517904 BC068166.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P517904 BC094570.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P517904 BC118503.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P517904 AK144349.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P517904 BC064727.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130955 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P141416 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P157507 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15093622 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15093626 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15093624 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15093627 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15093618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32707077 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32707044 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32707045 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32707039 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32707031 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32707056 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32707060 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32707064 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32707066 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32707071 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32707016 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32707062 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32707037 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32707023 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32707017 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32707026 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32707050 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32707054 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32707034 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32707068 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32707028 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14607811 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32707055 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32707046 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32707022 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32707069 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32707025 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32707032 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32707019 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14612115 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32707036 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32707059 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32707024 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32707070 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32707027 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14943313 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32707015 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32707058 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32707061 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32707030 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32707048 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32707018 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32707043 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32707035 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10602172 - -
          Affymetrix Mouse Gene 1.0 ST Array 10602171 - -
          Affymetrix Mouse Gene 1.0 ST Array 10602170 - -
          Affymetrix Mouse Gene 1.0 ST Array 10602168 - -
          Affymetrix Mouse Gene 1.0 ST Array 10602167 - -
          Affymetrix Mouse Gene 1.0 ST Array 10602169 - -
          Affymetrix HT MG-430 PM Array Plate 1456599_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10602166 - -
          Mouse Genome 430 2.0 Array 1456599_at - -
          Mouse Genome 430 2.0 Array 1456599_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 237082_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 237082_at - -
          Affymetrix Mouse Clariom S Array PSR0X00008896.mm.1 NM_001161430.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00008896.mm.1 NM_172782.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00008896.mm.1 TC0X00001402.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00008897.mm.1 NM_001161430.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00008897.mm.1 NM_172782.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00008897.mm.1 TC0X00001402.mm.1 MTA TC
          Mouse Expression Set 430 1456599_PM_at - -
          Mouse Expression Set 430 1456599_at - -
          Affymetrix HT Mouse Genome 430B Array 1456599_at - -
          Affymetrix HT Mouse Genome 430B Array 1456599_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2900008 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002900008 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1245658 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1245658 - -
          mouse-6 v1.1 (Illumina) scl0237082.4_163-S - -
          Illumina Sentrix mouse-6.v1 bead array 2900008 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0237082.4_163-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2900008 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1245658 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1245658 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0237082.4_163-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline