Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01332441_m1
          See other 1700007K13RIK GE Assays ›
          Gene Symbol:
          1700007K13Rik
          Gene Name:
          RIKEN cDNA 1700007K13 gene
          Gene Aliases:
          Rbest47
          Chromosome Location:
          Chr.2: 28462001 - 28466324 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01332441_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700007K13Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027040.1 NP_081316.1 2 - 3 365 95
          GenBank mRNA AK005731.1 - 2 - 3 375 95
          AY927869.1 - 2 - 3 310 95
          BC048581.1 - 2 - 3 346 95
          BC099566.1 - 2 - 3 365 30916995 95

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200003231.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1982559 NM_027040.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1982559 BC099566.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 107426 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 191946 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 97249 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1982559 NM_027040.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1982559 BC099566.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00330598 - -
          Whole Mouse Genome G4122A A_51_P169289 AK005731.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P30632 NM_027040.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P30632 BC048581.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P30632 BC099566.1 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01267084 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01267105 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01267088 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01204913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21091264 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01251162 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21091318 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01267104 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01401190 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01267094 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21091287 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21091298 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01267098 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02129342 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21091322 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02129334 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21091301 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01267100 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21091326 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01267085 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02129340 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21091282 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21091291 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02129337 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01267089 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01267101 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21091285 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21091283 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01251161 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21091284 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01267092 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21091297 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10481273 - -
          Affymetrix Mouse Gene 1.0 ST Array 10481277 - -
          Affymetrix Mouse Gene 1.0 ST Array 10481276 - -
          Affymetrix Mouse Gene 1.0 ST Array 10481274 - -
          Affymetrix Mouse Gene 1.0 ST Array 10481275 - -
          Affymetrix HT MG-430 PM Array Plate 1428705_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10481272 - -
          Mouse Genome 430 2.0 Array 1428705_PM_at - -
          Mouse Genome 430 2.0 Array 1428705_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69327_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69327_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0200027222.mm.1 NM_027040.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200027222.mm.1 NM_027040.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200027222.mm.1 TC0200003231.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200027223.mm.1 NM_027040.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200027223.mm.1 NM_027040.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200027223.mm.1 TC0200003231.mm.1 MTA TC
          Mouse Expression Set 430 1428705_at - -
          Mouse Expression Set 430 1428705_PM_at - -
          Murine Genome U74v2 Set 165968_f_at - -
          Murine Genome U74v2 Set 170283_at - -
          Murine Genome U74v2 Set 170283_PM_at - -
          Murine Genome U74v2 Set 165968_PM_f_at - -
          Affymetrix HT Mouse Genome 430B Array 1428705_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1428705_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 642048 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2480128 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1232336 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1232336 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline