Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01332972_m1
          See other ZFP777 GE Assays ›
          Gene Symbol:
          Zfp777
          Gene Name:
          zinc finger protein 777
          Gene Aliases:
          2500002G23Rik
          Chromosome Location:
          Chr.6: 48017660 - 48050236 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01332972_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp777
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001081382.1 NP_001074851.1 2 - 3 986 59
          GenBank mRNA BC066046.1 - 2 - 3 1076 59
          BC150871.1 - 2 - 3 1010 9087782 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600002216.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2002472 AK010853.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2002472 NM_001081382.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 320038 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 73871 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 396449 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 858511 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 252203 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 808604 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 616045 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 466017 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 660358 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 669854 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 357090 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 176446 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 403815 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 342188 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 596231 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 599403 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 60832 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 139811 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P120818 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P114699 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P115961 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P146811 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P152112 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P154768 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2002472 AK010853.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2002472 NM_001081382.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P446731 AK010853.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P446731 NM_001081382.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P159951 BC150871.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P159951 BC066046.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P159951 BC020288.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P159951 NM_001081382.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24425418 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05311501 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05052166 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24425485 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24425330 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05601153 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24425473 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24425332 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24425486 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05311503 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05727436 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05255844 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24425478 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24425368 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24425509 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24425363 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05208155 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05879297 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24425341 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24425467 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P04998309 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24425359 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24425354 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24425488 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24425549 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05343353 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24425482 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05827611 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24425517 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24425510 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P04998312 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24425357 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24425487 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24425506 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05827618 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24425471 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24425489 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24425336 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24425501 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24425481 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24425546 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24425551 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P04998327 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05083664 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24425503 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05066919 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24425540 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24425526 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24425537 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24425492 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05601143 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05186363 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24425365 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24425491 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24425339 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05601150 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24425361 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24425480 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24425490 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24425522 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24425465 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05311496 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24425545 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05208156 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05374724 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24425479 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24425369 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24425543 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24425464 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24425468 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24425519 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24425531 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10544554 - -
          Affymetrix Mouse Gene 1.0 ST Array 10544549 - -
          Affymetrix Mouse Gene 1.0 ST Array 10544550 - -
          Affymetrix Mouse Gene 1.0 ST Array 10544551 - -
          Affymetrix Mouse Gene 1.0 ST Array 10544552 - -
          Affymetrix Mouse Gene 1.0 ST Array 10544548 - -
          Affymetrix Mouse Gene 1.0 ST Array 10544553 - -
          Affymetrix HT MG-430 PM Array Plate 1429541_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10544547 - -
          Mouse Genome 430 2.0 Array 1429541_PM_at - -
          Mouse Genome 430 2.0 Array 1429541_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 72306_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 72306_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0600017412.mm.1 TC0600002216.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600017412.mm.1 NM_001081382.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017412.mm.1 NM_001081382.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017414.mm.1 TC0600002216.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600017414.mm.1 NM_001081382.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017414.mm.1 NM_001081382.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017416.mm.1 TC0600002216.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600017416.mm.1 NM_001081382.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017416.mm.1 NM_001081382.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017413.mm.1 TC0600002216.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600017413.mm.1 NM_001081382.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017413.mm.1 NM_001081382.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017418.mm.1 TC0600002216.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600017418.mm.1 NM_001081382.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017418.mm.1 NM_001081382.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017415.mm.1 TC0600002216.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600017415.mm.1 NM_001081382.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017415.mm.1 NM_001081382.1 Refseq
          Mouse Expression Set 430 1429541_PM_at - -
          Mouse Expression Set 430 1429541_at - -
          Murine Genome U74v2 Set 104353_at - -
          Murine Genome U74v2 Set 92664_at - -
          Murine Genome U74v2 Set 92664_PM_at - -
          Murine Genome U74v2 Set 104353_PM_at - -
          Mouse Genome 430A 2.0 Array 1429541_at - -
          Mouse Genome 430A 2.0 Array 1429541_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1429541_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1429541_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 316991 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1260244 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1243697 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2897328 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2897328 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0