Search
Search

Pmp22| Gene Symbol: Pmp22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK076005.1 | - | 2 - 3 | 212 | 72 | |
| AK171745.1 | - | 2 - 3 | 209 | 72 | ||
| BC010765.1 | - | 2 - 3 | 235 | 4225316 | 72 | |
| BY092580.1 | - | 2 - 3 | 318 | 72 | ||
| BY234063.1 | - | 2 - 3 | 270 | 72 | ||
| BY235328.1 | - | 2 - 3 | 262 | 72 | ||
| BY235583.1 | - | 2 - 3 | 288 | 72 | ||
| Z38110.1 | - | 1 - 2 | 76 | 72 | ||
Set Membership: |
|