Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01333405_m1
          See other PIP5KL1 GE Assays ›
          Gene Symbol:
          Pip5kl1
          Gene Name:
          phosphatidylinositol-4-phosphate 5-kinase-like 1
          Gene Aliases:
          BC028795
          Chromosome Location:
          Chr.2: 32574788 - 32583780 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01333405_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pip5kl1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_198191.2 NP_937834.1 9 - 10 960 64
          GenBank mRNA AK133244.1 - 9 - 10 2368 64
          AY376879.1 - 9 - 10 999 64
          BC028795.1 - 2 - 3 272 64
          BC053963.1 - 2 - 3 281 64
          BC117018.1 - 9 - 10 952 40126337 64
          BC119037.1 - 9 - 10 952 8733786 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200005470.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pip5kl1

          Panther Classification:

          Molecular Function -

          kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          phosphorylation
          phosphatidylinositol metabolic process
          phosphatidylinositol phosphorylation
          nucleotide binding
          protein binding
          ATP binding
          kinase activity
          phosphatidylinositol phosphate kinase activity
          1-phosphatidylinositol-4-phosphate 5-kinase activity
          transferase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21116214 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21116215 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2002309 BC094346.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2002309 AK164998.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2002309 AY376879.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2002309 BC028795.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2002309 BC053963.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 396094 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 670234 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 851059 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 940686 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 71410 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 94421 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P161901 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2002309 BC094346.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2002309 AK164998.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2002309 AY376879.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2002309 BC028795.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2002309 BC053963.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21116218 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21116168 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21116166 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21116217 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21116216 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21116164 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21116167 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21116210 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21116171 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21116163 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21116208 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21116176 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21116159 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21116160 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21116214 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21116157 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21116162 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21116161 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21116207 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21116211 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21116173 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21116170 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P161901 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04476836 - -
          Whole Mouse Genome G4122A A_51_P331798 BC028795.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P331798 BC053963.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123342 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100739 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128147 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P161901 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01613682 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01613683 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01731731 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01731738 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01731730 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01731729 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01906451 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01906448 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21116215 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21116214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21116171 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01785938 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21116163 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21116171 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21116207 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01785937 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01715822 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21116211 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01424474 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21116179 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21116167 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21116151 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21116161 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21116182 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01942004 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21116183 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21116160 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21116176 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01293020 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01500610 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01715819 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21116186 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21116173 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21116218 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21116154 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01785942 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21116216 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21116208 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21116150 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21116146 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21116214 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21116178 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01605031 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21116162 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21116148 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21116197 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21116155 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21116157 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21116145 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21116201 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21116198 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21116210 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21116217 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01701119 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01330633 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21116190 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21116200 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10471450 - -
          Affymetrix Mouse Gene 1.0 ST Array 10471446 - -
          Affymetrix Mouse Gene 1.0 ST Array 10471456 - -
          Affymetrix Mouse Gene 1.0 ST Array 10471449 - -
          Affymetrix Mouse Gene 1.0 ST Array 10471451 - -
          Affymetrix Mouse Gene 1.0 ST Array 10471452 - -
          Affymetrix Mouse Gene 1.0 ST Array 10471454 - -
          Affymetrix Mouse Gene 1.0 ST Array 10471455 - -
          Affymetrix Mouse Gene 1.0 ST Array 10471448 - -
          Affymetrix Mouse Gene 1.0 ST Array 10471447 - -
          Affymetrix Mouse Gene 1.0 ST Array 10471444 - -
          Affymetrix Mouse Gene 1.0 ST Array 10471453 - -
          Affymetrix HT MG-430 PM Array Plate 1427900_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10471443 - -
          Mouse Genome 430 2.0 Array 1427900_PM_at - -
          Mouse Genome 430 2.0 Array 1427900_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 227733_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 227733_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0200005299.mm.1 NM_198191.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005299.mm.1 TC0200005470.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200005301.mm.1 NM_198191.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005301.mm.1 TC0200005470.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200005300.mm.1 NM_198191.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005300.mm.1 TC0200005470.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200005298.mm.1 NM_198191.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005298.mm.1 TC0200005470.mm.1 MTA TC
          Mouse Expression Set 430 1427900_at - -
          Mouse Expression Set 430 1427900_PM_at - -
          Murine Genome U74v2 Set 166245_PM_at - -
          Murine Genome U74v2 Set 166245_at - -
          Mouse Genome 430A 2.0 Array 1427900_PM_at - -
          Mouse Genome 430A 2.0 Array 1427900_at - -
          Affymetrix HT Mouse Genome 430A Array 1427900_at - -
          Affymetrix HT Mouse Genome 430A Array 1427900_PM_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1410577 - -
          Illumina mouse-6 v1.0 expression beadchip 1410577 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001410577 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1223829 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1223829 - -
          mouse-6 v1.1 (Illumina) scl21057.3.1_51-S - -
          Illumina Sentrix mouse-6.v1 bead array 1410577 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl21057.3.1_51-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1410577 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1223829 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1223829 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline