Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01333562_m1
          See other ANKRD16 GE Assays ›
          Gene Symbol:
          Ankrd16
          Gene Name:
          ankyrin repeat domain 16
          Gene Aliases:
          2810455F06Rik, AI646698, D430029B21Rik
          Chromosome Location:
          Chr.2: 11777753 - 11790327 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01333562_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ankrd16
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_177268.4 NP_796242.2 6 - 7 1763 60
          GenBank mRNA AK013354.1 - 3 - 4 701 60
          AK140039.1 - 5 - 6 3118 60
          BC021802.1 - 6 - 7 892 60
          BC037442.1 - 5 - 6 3047 60
          BC065408.1 - 2 - 3 326 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200000223.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ankrd16

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21007914 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21007915 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P127175 BC037442.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P127175 BC065408.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P127175 AK140039.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P127175 BC021802.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P127175 NM_177268.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2075333 BC116229.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2075333 BC116230.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2075333 AK052460.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2075333 AK085853.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 222747 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 784465 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 456947 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 672505 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 451872 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 146178 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 704947 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 495858 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 57825 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P150091 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2075333 BC116229.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2075333 BC116230.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2075333 AK052460.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2075333 AK085853.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P127175 BC037442.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P127175 BC065408.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P127175 AK140039.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P127175 BC021802.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P127175 NM_177268.4 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21007937 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21007923 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21007930 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21007943 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21007924 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21007938 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21007918 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21007914 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21007932 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21007935 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21007922 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21007915 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21007925 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21007940 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21007939 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21007916 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21007921 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21007942 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21007933 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21007926 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21007920 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04430181 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P150091 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00305696 - -
          Whole Mouse Genome G4122A A_52_P265773 AK013354.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P332642 AK052460.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127175 BC037442.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127175 BC065408.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127175 AK140039.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127175 BC021802.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127175 NM_177268.4 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P150091 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01314387 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21007914 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21007915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21007844 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21008009 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21007976 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21007963 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21007940 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21007920 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21007953 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21007986 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21007971 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21007910 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01591267 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21007955 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21007942 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21007918 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21007932 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21008005 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01680996 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21007925 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21007957 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21008011 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21007913 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21008014 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21007937 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01591262 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21007949 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01314387 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21008003 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21007944 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21007930 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21007961 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21007989 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21007912 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01367137 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01935807 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21007983 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01314374 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21007954 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21007939 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21007990 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21007973 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21007933 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21007916 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21008016 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21007982 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21007946 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21007922 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01262418 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01194423 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01314392 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21007958 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21007938 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01275186 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21007966 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21008012 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21007935 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21007959 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10469304 - -
          Affymetrix Mouse Gene 1.0 ST Array 10469307 - -
          Affymetrix Mouse Gene 1.0 ST Array 10469301 - -
          Affymetrix Mouse Gene 1.0 ST Array 10469303 - -
          Affymetrix Mouse Gene 1.0 ST Array 10469302 - -
          Affymetrix Mouse Gene 1.0 ST Array 10469310 - -
          Affymetrix Mouse Gene 1.0 ST Array 10469311 - -
          Affymetrix Mouse Gene 1.0 ST Array 10469308 - -
          Affymetrix Mouse Gene 1.0 ST Array 10469309 - -
          Affymetrix Mouse Gene 1.0 ST Array 10469306 - -
          Affymetrix HT MG-430 PM Array Plate 1428239_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10469300 - -
          Mouse Genome 430 2.0 Array 1428239_at - -
          Mouse Genome 430 2.0 Array 1428239_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0200000896.mm.1 NM_177268.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200000896.mm.1 NM_177268.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200000896.mm.1 TC0200000223.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200000898.mm.1 NM_177268.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200000898.mm.1 NM_177268.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200000898.mm.1 TC0200000223.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200000897.mm.1 NM_177268.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200000897.mm.1 NM_177268.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200000897.mm.1 TC0200000223.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200000899.mm.1 NM_177268.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200000899.mm.1 NM_177268.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200000899.mm.1 TC0200000223.mm.1 MTA TC
          Mouse Expression Set 430 1428239_PM_at - -
          Mouse Expression Set 430 1428239_at - -
          Affymetrix HT Mouse Genome 430B Array 1428239_at - -
          Affymetrix HT Mouse Genome 430B Array 1428239_PM_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3870411 - -
          Illumina mouse-6 v1.0 expression beadchip 3870411 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003870411 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1258280 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1258280 - -
          mouse-6 v1.1 (Illumina) scl00320816.1_329-S - -
          Illumina Sentrix mouse-6.v1 bead array 3870411 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00320816.1_329-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3870411 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1258280 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1258280 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline