Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01333971_m1
          See other GPCPD1 GE Assays ›
          Gene Symbol:
          Gpcpd1
          Gene Name:
          glycerophosphocholine phosphodiesterase GDE1 homolog (S. cerevisiae)
          Gene Aliases:
          2310004G06Rik, 2310032D16Rik, AU015213, Gde5, Prei4, mKIAA1434
          Chromosome Location:
          Chr.2: 132529082 - 132578282 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01333971_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gpcpd1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001042672.1 NP_001036137.1 8 - 9 919 86
          NM_001291050.1 NP_001277979.1 7 - 8 792 86
          NM_001291051.1 NP_001277980.1 7 - 8 822 86
          NM_001291052.1 NP_001277981.1 7 - 8 840 86
          NM_027096.2 NP_081372.1 6 - 7 716 86
          NM_028802.3 NP_083078.3 8 - 9 916 86
          GenBank mRNA AK009563.1 - 1 - 2 135 86
          AK030645.1 - 8 - 9 870 86
          AK031658.1 - 7 - 8 795 86
          AK050318.1 - 8 - 9 831 86
          AK141374.1 - 8 - 9 805 86
          AK163419.1 - 7 - 8 762 86
          AK165952.1 - 8 - 9 881 86
          AK166454.1 - 7 - 8 805 86
          BC006887.1 - 7 - 8 764 86
          BC033408.1 - 6 - 7 646 5357683 86

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AK009563.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gpcpd1

          Panther Classification:

          Molecular Function -

          hydrolase phosphodiesterase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid metabolic process
          skeletal muscle tissue development
          phosphoric diester hydrolase activity
          hydrolase activity
          carbohydrate binding
          glycerophosphocholine phosphodiesterase activity
          starch binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200040173.mm.1 NM_001042672.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200040173.mm.1 NM_001042672.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200040173.mm.1 NM_028802.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200040173.mm.1 TC0200004676.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200040174.mm.1 NM_001042672.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200040174.mm.1 NM_001042672.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200040174.mm.1 NM_028802.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200040174.mm.1 TC0200004676.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline