Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01334182_mH
          See other ZFP120 GE Assays ›
          Gene Symbol:
          Zfp120
          Gene Name:
          zinc finger protein 120
          Gene Aliases:
          AI315103, E030042N05Rik, F80, MZF31, Zfp844
          Chromosome Location:
          Chr.2: 150114407 - 150136678 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01334182_mH
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp120
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_181266.2 NP_851783.2 3 - 4 297 160
          GenBank mRNA AF211867.1 - 3 - 4 297 160
          AK033399.1 - 3 - 4 294 160
          AK132318.1 - 3 - 4 303 160
          BC013500.1 - 3 - 4 300 160
          BC141128.1 - 3 - 4 287 9055740 160

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200004826.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zfp120

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          biological_process
          nucleic acid binding
          DNA binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P476261 AF211869.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P476261 AF211867.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P476261 AF211868.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P476261 AF211869.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P476261 AF211867.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P476261 AF211868.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2136672 BC141128.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2136672 AK132318.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2136672 BC013500.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2136672 AF211869.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2136672 AF242378.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 73586 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 426569 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 177920 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 373339 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 527513 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 429554 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 175645 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 271136 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 690763 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 11442 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 180117 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 12929 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2136672 BC141128.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2136672 AK132318.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2136672 BC013500.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2136672 AF211869.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2136672 AF242378.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P476261 AF211869.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P476261 AF211867.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P476261 AF211868.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P476261 AF211869.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P476261 AF211867.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P476261 AF211868.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21738437 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21738401 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21738428 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21738416 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00525859 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04736975 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04736972 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00525869 - -
          Whole Mouse Genome G4122A A_51_P476261 AF211869.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P476261 AF211867.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P476261 AF211868.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P476261 AF211869.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P476261 AF211867.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P476261 AF211868.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P00525859 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P00525869 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01894619 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01894614 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01894615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21738392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21738315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21738389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21738323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21738371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21738353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21738405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21738325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21738357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21738349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21738369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21738346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21738378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21738375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21738358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21738311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21738398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21738342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21738329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21738363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21738351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21738361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21738354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21738356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21738333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21738360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21738377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21738324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21738352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21738313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21738328 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21738378 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21738380 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21738392 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01729778 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21738437 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02091060 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21738390 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01729767 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21738428 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21738398 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21738389 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21738416 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01894615 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01894618 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21738405 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21738422 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21738440 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21738387 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21738401 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10488448 - -
          Affymetrix Mouse Gene 1.0 ST Array 10488442 - -
          Affymetrix Mouse Gene 1.0 ST Array 10488444 - -
          Affymetrix Mouse Gene 1.0 ST Array 10488446 - -
          Affymetrix Mouse Gene 1.0 ST Array 10488443 - -
          Affymetrix Mouse Gene 1.0 ST Array 10488447 - -
          Affymetrix Mouse Gene 1.0 ST Array 10488445 - -
          Affymetrix HT MG-430 PM Array Plate 1457433_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1421519_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10488441 - -
          Mouse Genome 430 2.0 Array 1421519_PM_a_at - -
          Mouse Genome 430 2.0 Array 1421519_a_at - -
          Mouse Genome 430 2.0 Array 1457433_x_at - -
          Mouse Genome 430 2.0 Array 1457433_PM_x_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 104348_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 104348_at - -
          Affymetrix Mouse Clariom S Array PSR0200041144.mm.1 NM_023266.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041144.mm.1 NM_023266.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041144.mm.1 NM_181266.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041144.mm.1 NM_181266.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041144.mm.1 TC0200004826.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200041140.mm.1 NM_023266.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041140.mm.1 NM_023266.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041140.mm.1 NM_181266.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041140.mm.1 NM_181266.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041140.mm.1 TC0200004826.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200041141.mm.1 NM_023266.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041141.mm.1 NM_023266.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041141.mm.1 NM_181266.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041141.mm.1 NM_181266.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041141.mm.1 TC0200004826.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200041142.mm.1 NM_023266.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041142.mm.1 NM_023266.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041142.mm.1 NM_181266.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041142.mm.1 NM_181266.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041142.mm.1 TC0200004826.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200041145.mm.1 NM_023266.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041145.mm.1 NM_023266.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041145.mm.1 NM_181266.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041145.mm.1 NM_181266.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041145.mm.1 TC0200004826.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200041139.mm.1 NM_023266.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041139.mm.1 NM_023266.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041139.mm.1 NM_181266.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041139.mm.1 NM_181266.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041139.mm.1 TC0200004826.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200041148.mm.1 NM_181266.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041148.mm.1 NM_181266.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200041148.mm.1 TC0200004826.mm.1 MTA TC
          Mouse Expression Set 430 1421519_PM_a_at - -
          Mouse Expression Set 430 1421519_a_at - -
          Mouse Expression Set 430 1457433_PM_x_at - -
          Mouse Expression Set 430 1457433_x_at - -
          Mouse Genome 430A 2.0 Array 1421519_a_at - -
          Mouse Genome 430A 2.0 Array 1421519_PM_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1457433_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1457433_PM_x_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3710279 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2456317 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3065805 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3065805 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline