Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01334204_s1
          See other OLFR1155 GE Assays ›
          Gene Symbol:
          Olfr1155
          Gene Name:
          olfactory receptor 1155
          Gene Aliases:
          MOR174-10
          Chromosome Location:
          Chr.2: 87942682 - 87943626 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01334204_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr1155
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146643.2 NP_666854.1 1 552 112

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr1155

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2085531 NM_146643.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 579888 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P170788 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2085531 NM_146643.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P63637 NM_146643.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P170788 - -
          Whole Mouse Genome G4122A A_52_P63637 NM_146643.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100581 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P170788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21407867 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407871 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407854 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02161733 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407880 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407878 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407849 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407867 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407881 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407840 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407862 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407859 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02107465 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407842 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407865 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407877 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407870 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407874 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02161768 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407860 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407848 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407851 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407869 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407873 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407856 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407858 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407846 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21407844 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10484700 - -
          Affymetrix Mouse Gene 1.1 ST Array 10484699 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 568871 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4570687 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004570687 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1226210 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1226210 - -
          mouse-6 v1.1 (Illumina) scl19040.1.1_9-S - -
          Illumina Sentrix mouse-6.v1 bead array 4570687 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl19040.1.1_9-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4570687 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1226210 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1226210 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0