| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28958205 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28958207 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28958208 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28958204 |
- |
- |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2034335 |
AY695827.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2185100 |
BC082568.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
366937 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
346803 |
AK013901.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
346803 |
BC003342.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
346803 |
NM_008894.1 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
346803 |
Z72486.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
281576 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
479885 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
480587 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
518322 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
435068 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
239372 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
216632 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
60213 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
551261 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
379317 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
726181 |
AK031458.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
726181 |
AK081382.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
726181 |
BC046779.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
726181 |
BC060735.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
726181 |
XM_001476544.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
726181 |
XM_001481308.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
853440 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
148134 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
342867 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
968071 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
656704 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
493847 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
36937 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
682074 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
594889 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
237936 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
71037 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
71405 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
158211 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
128422 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
94062 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
174276 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
25744 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
66667 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P155695 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2034335 |
AY695827.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2185100 |
BC082568.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P116919 |
AY695827.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P116919 |
AK132733.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P116919 |
NM_027724.2 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28958197 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28958196 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28958191 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28958169 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28958202 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28958200 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28958189 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28958194 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28958180 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28958170 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28958205 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28958174 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28958185 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28958206 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28958177 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28958192 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28958199 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28958172 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28958182 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28958187 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28958201 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28958176 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P02668202 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P155695 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07841805 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P02668213 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07841754 |
- |
- |
| Whole Mouse Genome G4122A |
A_52_P116919 |
AY695827.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P116919 |
AK132733.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P116919 |
NM_027724.2 |
mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P155695 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P02668213 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10693265 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10693260 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10693264 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10693259 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28958204 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28958208 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28958207 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28958205 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958068 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958202 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958050 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958000 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958120 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958024 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958047 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958054 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958094 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958086 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958127 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958090 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958144 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958079 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958218 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958226 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958002 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958210 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958237 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28957999 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958131 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958026 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958165 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958142 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958209 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958062 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958216 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958212 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958039 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958007 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958081 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958096 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958171 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958197 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958055 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958123 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958029 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958005 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958176 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958117 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958124 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958140 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958150 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958083 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28957994 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958133 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958228 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958063 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958052 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958023 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958138 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958058 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958066 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958012 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958233 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958109 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958065 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958121 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958191 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958118 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958230 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958180 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958021 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958070 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958183 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958186 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958004 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958041 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958088 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958134 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958017 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958049 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958110 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958143 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958206 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958214 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958129 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958113 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958102 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958082 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958042 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958137 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958057 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28957995 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958006 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958077 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958107 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958148 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958238 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958033 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958092 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958103 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958231 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958147 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28957991 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958201 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958158 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958205 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958037 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958199 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958185 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958111 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958115 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958178 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958223 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958136 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958200 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958043 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958020 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958035 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958161 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958059 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958220 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958154 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958084 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28958125 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28958143 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28958177 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28958168 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28958163 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28958189 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28958152 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28958154 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28958169 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28958178 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28958165 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28958156 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P10719619 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28958164 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28958144 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28958191 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28958174 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28958180 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28958147 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P10711627 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28958183 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28958176 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28958142 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P10792054 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28958158 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28958148 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28958185 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P11087893 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28958170 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P10774879 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28958161 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28958194 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P28958175 |
- |
- |