| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1700007412.mm.1 |
NR_033517.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700007412.mm.1 |
NR_033517.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700007412.mm.1 |
TC1700000722.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700007417.mm.1 |
NR_033517.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700007417.mm.1 |
NR_033517.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700007417.mm.1 |
TC1700000722.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700007416.mm.1 |
NR_033517.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700007416.mm.1 |
NR_033517.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700007416.mm.1 |
TC1700000722.mm.1 |
MTA TC |