| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200006294.mm.1 |
NM_029687.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200006294.mm.1 |
NM_029687.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200006294.mm.1 |
TC0200000718.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200006295.mm.1 |
NM_029687.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200006295.mm.1 |
NM_029687.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200006295.mm.1 |
TC0200000718.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200006293.mm.1 |
NM_029687.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200006293.mm.1 |
NM_029687.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200006293.mm.1 |
TC0200000718.mm.1 |
MTA TC |