Search
Search

Zfp865| Gene Symbol: Zfp865 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290426.1 | NP_001277355.1 | 2 - 3 | 2571 | 103 | |
| GenBank mRNA | AK051004.1 | - | 2 - 3 | 2377 | 103 | |
Set Membership: |
|