Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01335566_g1
          See other BC051665 GE Assays ›
          Gene Symbol:
          BC051665
          Gene Name:
          cDNA sequence BC051665
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 60781887 - 60786364 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01335566_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: BC051665
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_199148.2 NP_954599.2 6 - 7 869 72
          GenBank mRNA AK145559.1 - 6 - 7 867 72
          BC051665.1 - 6 - 7 854 30027545 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300002138.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Protease 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene BC051665

          Panther Classification:

          Molecular Function -

          cysteine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          proteolysis involved in cellular protein catabolic process
          molecular_function
          cysteine-type endopeptidase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2008913 BC051665.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2008913 NM_199148.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2008913 AK145559.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 409101 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 872223 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 810807 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P150668 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2008913 BC051665.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2008913 NM_199148.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2008913 AK145559.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087531 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087530 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087519 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087544 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087546 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087539 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087541 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087512 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087548 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087542 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087550 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087537 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087535 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087525 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087547 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087509 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087523 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087526 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087551 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087527 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087553 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29087534 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P150668 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07896235 - -
          Whole Mouse Genome G4122A A_52_P605319 BC051665.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P605319 NM_199148.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P605319 AK145559.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133207 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P150668 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10551110 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10551103 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10551100 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10551115 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10551098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29087486 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087437 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087537 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11107916 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087544 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087482 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087550 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087553 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087542 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087527 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087514 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087557 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087508 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087551 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087495 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087433 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087523 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087497 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087525 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087558 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087481 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087493 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087512 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087483 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087548 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087486 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087555 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10832156 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087521 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087509 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11097020 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087435 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087526 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10551104 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087473 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087531 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087530 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087547 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087491 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10832149 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11085833 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087545 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10897464 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087546 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11097030 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087472 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087484 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087541 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087534 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10897463 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087506 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087476 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087503 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087539 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11097016 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087501 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29087556 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10551106 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10409844 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409846 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409843 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409842 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409845 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409847 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409841 - -
          Affymetrix Mouse Gene 1.1 ST Array 10409840 - -
          Affymetrix Mouse Clariom S Array PSR1300013708.mm.1 NM_199148.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013708.mm.1 NM_199148.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013708.mm.1 TC1300002138.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300013707.mm.1 NM_199148.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013707.mm.1 NM_199148.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300013707.mm.1 TC1300002138.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 383271 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4200154 - -
          Illumina mouse-6 v1.0 expression beadchip 4200154 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004200154 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1221521 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1221521 - -
          mouse-6 v1.1 (Illumina) scl0218275.1_67-S - -
          Illumina Sentrix mouse-6.v1 bead array 4200154 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0218275.1_67-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4200154 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2910076 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2910076 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1221521 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline