| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100007254.mm.1 |
NM_001004435.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100007254.mm.1 |
NM_001081566.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100007254.mm.1 |
TC1100000873.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100007256.mm.1 |
NM_001004435.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100007256.mm.1 |
NM_001081566.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100007256.mm.1 |
TC1100000873.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100007255.mm.1 |
NM_001004435.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100007255.mm.1 |
NM_001081566.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100007255.mm.1 |
TC1100000873.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100007257.mm.1 |
NM_001004435.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100007257.mm.1 |
NM_001081566.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100007257.mm.1 |
TC1100000873.mm.1 |
MTA TC |