Search
Search

Fbln7| Gene Symbol: Fbln7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024237.4 | NP_077199.2 | 3 - 4 | 548 | 77 | |
| GenBank mRNA | AK005465.1 | - | 3 - 4 | 503 | 77 | |
| BC051550.1 | - | 1 - 2 | 104 | 77 | ||
| BC095941.1 | - | 3 - 4 | 480 | 4167912 | 77 | |
| CK023294.1 | - | 3 - 4 | 490 | 77 | ||
| EF668007.1 | - | 3 - 4 | 407 | 77 | ||
Set Membership: |
|