Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01336295_m1
          See other CELF2 GE Assays ›
          Gene Symbol:
          Celf2
          Gene Name:
          CUGBP, Elav-like family member 2
          Gene Aliases:
          B230218O03, B230345P09Rik, C88023, CELF-2, CUG-BP2, Cugbp2, D230046B21Rik, Etr-3, Napor, Napor-2, mETR-3
          Chromosome Location:
          Chr.2: 6539694 - 7396227 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01336295_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Celf2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001110228.1 NP_001103698.1 8 - 9 914 65
          NM_001110229.1 NP_001103699.1 9 - 10 1001 65
          NM_001110230.1 NP_001103700.1 9 - 10 1001 65
          NM_001110231.1 NP_001103701.1 11 - 12 976 65
          NM_001110232.1 NP_001103702.1 10 - 11 987 65
          NM_001160292.1 NP_001153764.1 9 - 10 1001 65
          NM_001160293.1 NP_001153765.1 8 - 9 914 65
          NM_010160.2 NP_034290.2 8 - 9 914 65
          GenBank mRNA AF090696.1 - 10 - 11 987 65
          AF090697.1 - 8 - 9 887 65
          AK135166.1 - 8 - 9 866 65
          AK137292.1 - 9 - 10 942 65
          AK151818.1 - 7 - 8 772 65
          AK152628.1 - 7 - 8 772 65
          AK153267.1 - 7 - 8 772 65
          AK159996.1 - 9 - 10 1006 65
          AK160861.1 - 11 - 12 977 65
          AK162651.1 - 6 - 7 4477 65
          AK164914.1 - 9 - 10 942 65
          BC026856.1 - 8 - 9 865 4503295 65
          Y18298.1 - 7 - 8 780 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200002894.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Celf2

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA splice site selection
          mRNA processing
          nucleotide binding
          nucleic acid binding
          RNA binding
          poly(A) RNA binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200024062.mm.1 NM_001110228.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200024062.mm.1 NM_001110228.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200024062.mm.1 NM_001110229.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200024062.mm.1 NM_001110229.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200024062.mm.1 NM_001110230.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200024062.mm.1 NM_001110230.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200024062.mm.1 NM_001110231.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200024062.mm.1 NM_001110231.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200024062.mm.1 NM_001110232.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200024062.mm.1 NM_001110232.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200024062.mm.1 NM_001160292.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200024062.mm.1 NM_001160292.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200024062.mm.1 NM_001160293.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200024062.mm.1 NM_001160293.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200024062.mm.1 NM_010160.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200024062.mm.1 NM_010160.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200024062.mm.1 TC0200002894.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline