Search
Search

Tnks1bp1| Gene Symbol: Tnks1bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081260.2 | NP_001074729.1 | 9 - 10 | 5068 | 91 | |
| GenBank mRNA | AK081682.1 | - | 1 - 2 | 1422 | 91 | |
| AK129436.1 | - | 5 - 6 | 3761 | 91 | ||
| BC024499.1 | - | 4 - 5 | 966 | 91 | ||
| BC025657.1 | - | 2 - 3 | 547 | 91 | ||
| BC025943.1 | - | 4 - 5 | 2517 | 91 | ||
| BC058639.1 | - | 6 - 7 | 4212 | 91 | ||
| BC150947.1 | - | 9 - 10 | 4988 | 9087858 | 91 | |
Set Membership: |
|