Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01336731_m1
          See other A230065H16RIK GE Assays ›
          Gene Symbol:
          A230065H16Rik
          Gene Name:
          RIKEN cDNA A230065H16 gene
          Gene Aliases:
          Gm898, Lbh2
          Chromosome Location:
          Chr.12: 111402047 - 111412077 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01336731_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: A230065H16Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001101503.1 NP_001094973.1 3 - 4 306 76
          GenBank mRNA AK038816.1 - 3 - 4 306 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200001235.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Recommended Assay Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2076916 NM_001101503.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2076916 AK038816.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 227234 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 420335 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 434827 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2076916 NM_001101503.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2076916 AK038816.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28743803 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28743864 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28743849 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28743815 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10322519 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28743817 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10008196 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28743871 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28743882 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28743839 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28743855 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10132490 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09812066 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28743842 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28743807 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28743865 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28743873 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28743833 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28743831 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10322516 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28743834 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09987395 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28743867 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09829351 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09829338 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28743879 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10309006 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28743892 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28743887 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10295696 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10082869 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28743889 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28743844 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28743888 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28743806 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09829327 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28743809 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28743877 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09829335 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28743828 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28743825 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28743816 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28743891 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10322520 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09829348 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28743893 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10322523 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28743810 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28743858 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28743884 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28743840 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P09812071 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28743866 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28743886 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28743872 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28743823 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1442011_PM_at - -
          Mouse Genome 430 2.0 Array 1442011_PM_at - -
          Mouse Genome 430 2.0 Array 1442011_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 380787_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 380787_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1200008719.mm.1 NM_001101503.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008719.mm.1 NM_001101503.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008719.mm.1 TC1200001235.mm.1 MTA TC
          Affymetrix HT Mouse Genome 430B Array 1442011_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1442011_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 510075 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1222045 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline