Search
Search

4930402F06Rik| Gene Symbol: 4930402F06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080709.2 | NP_001074178.1 | 4 - 5 | 441 | 63 | |
| NM_001272040.1 | NP_001258969.1 | 3 - 4 | 357 | 63 | ||
| GenBank mRNA | AK015047.1 | - | 4 - 5 | 442 | 63 | |
| BC115676.1 | - | 3 - 4 | 240 | 40104161 | 63 | |
| BC115677.1 | - | 3 - 4 | 240 | 40104162 | 63 | |
Set Membership: |
|