Search
Search

Gapvd1| Gene Symbol: Gapvd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025709.2 | NP_079985.2 | 20 - 21 | 3809 | 60 | |
| GenBank mRNA | AK014474.1 | - | 5 - 6 | 675 | 60 | |
| AK088851.1 | - | 4 - 5 | 372 | 60 | ||
| AK129381.1 | - | 19 - 20 | 3404 | 60 | ||
| BC057164.1 | - | 5 - 6 | 678 | 60 | ||
| BC060123.1 | - | 20 - 21 | 3809 | 6408778 | 60 | |
| EF155419.1 | - | 18 - 19 | 3227 | 60 | ||
Set Membership: |
|