Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01336935_m1
          See other 1700026L06RIK GE Assays ›
          Gene Symbol:
          1700026L06Rik
          Gene Name:
          RIKEN cDNA 1700026L06 gene
          Gene Aliases:
          MAST
          Chromosome Location:
          Chr.2: 28692080 - 28699651 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01336935_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700026L06Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027283.1 NP_081559.1 597 61
          GenBank mRNA AK155408.1 - 4 - 5 598 61
          BC055108.1 - 597 6775232 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200003235.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene 1700026L06Rik

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          calcium-dependent protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21092718 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21092719 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21092715 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21092717 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21092716 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1979997 AK155297.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1979997 AK155408.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1979997 BC055108.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1979997 NM_027283.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1979998 AK155297.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1979998 BC055108.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1979998 NM_027283.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 151693 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 821783 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 500734 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 267990 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 728561 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 40117 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P179762 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1979997 AK155297.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1979997 AK155408.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1979997 BC055108.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1979997 NM_027283.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1979998 AK155297.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1979998 BC055108.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1979998 NM_027283.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21092682 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21092714 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21092680 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21092694 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21092700 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21092687 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21092697 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21092705 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21092717 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21092707 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21092702 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21092690 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21092685 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21092713 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21092709 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21092678 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21092684 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21092710 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21092698 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21092712 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21092718 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21092689 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21092692 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21092679 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21092695 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21092681 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00331014 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P179762 - -
          Whole Mouse Genome G4122A A_51_P513979 AK006393.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P179762 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01744570 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01364680 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01364679 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21092715 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21092717 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21092716 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21092719 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21092718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21092669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21092713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21092660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21092712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21092685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21092694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21092698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21092714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21092682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21092679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21092705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21092689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21092692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21092661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21092656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21092697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21092718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21092678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21092701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21092671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21092707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21092662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21092673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21092663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21092652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21092680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21092717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21092654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21092674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21092665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21092695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21092659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21092677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21092666 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21092692 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21092673 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21092701 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21092655 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21092661 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21092682 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21092689 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01892382 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21092671 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21092697 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21092665 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21092666 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21092709 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21092662 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01892381 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21092714 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21092659 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21092685 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01542972 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21092680 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21092717 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21092694 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21092695 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21092656 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01364676 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21092710 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01542997 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01542966 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01920143 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21092705 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21092660 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01542962 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21092674 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21092713 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21092707 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21092684 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10481314 - -
          Affymetrix Mouse Gene 1.0 ST Array 10481317 - -
          Affymetrix Mouse Gene 1.0 ST Array 10481319 - -
          Affymetrix Mouse Gene 1.0 ST Array 10481316 - -
          Affymetrix Mouse Gene 1.0 ST Array 10481313 - -
          Affymetrix Mouse Gene 1.0 ST Array 10481315 - -
          Affymetrix Mouse Gene 1.0 ST Array 10481318 - -
          Affymetrix HT MG-430 PM Array Plate 1452904_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10481312 - -
          Mouse Genome 430 2.0 Array 1452904_at - -
          Mouse Genome 430 2.0 Array 1452904_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69987_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69987_at - -
          Affymetrix Mouse Clariom S Array PSR0200027297.mm.1 NM_027283.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200027297.mm.1 NM_027283.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200027297.mm.1 TC0200003235.mm.1 MTA TC
          Mouse Expression Set 430 1452904_PM_at - -
          Mouse Expression Set 430 1452904_at - -
          Murine Genome U74v2 Set 171272_r_at - -
          Murine Genome U74v2 Set 169490_PM_r_at - -
          Murine Genome U74v2 Set 170450_PM_r_at - -
          Murine Genome U74v2 Set 170450_r_at - -
          Murine Genome U74v2 Set 171272_PM_r_at - -
          Murine Genome U74v2 Set 169490_r_at - -
          Affymetrix HT Mouse Genome 430B Array 1452904_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1452904_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 735232 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2720571 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2720571 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2720572 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline