Search
Search

Zfp773| Gene Symbol: Zfp773 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029584.1 | NP_083860.1 | 1 - 2 | 326 | 89 | |
| GenBank mRNA | AK013056.1 | - | 1 - 2 | 294 | 89 | |
| AK031040.1 | - | 1 - 2 | 326 | 89 | ||
| AK036639.1 | - | 1 - 2 | 296 | 89 | ||
| BC117497.1 | - | 1 - 2 | 304 | 40106595 | 89 | |
Set Membership: |
|