Search
Search

Arl10| Gene Symbol: Arl10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019968.2 | NP_064352.2 | 2 - 3 | 471 | 60 | |
| GenBank mRNA | AK205970.1 | - | 1 - 2 | 75 | 60 | |
| BC115498.2 | - | 2 - 3 | 412 | 40100921 | 60 | |
Set Membership: |
|