Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1300013168.mm.1 |
NM_001159513.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013168.mm.1 |
NM_001159513.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013168.mm.1 |
NM_001159514.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013168.mm.1 |
NM_001159514.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013168.mm.1 |
NM_001159515.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013168.mm.1 |
NM_001159515.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013168.mm.1 |
NM_012015.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013168.mm.1 |
NM_012015.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013168.mm.1 |
TC1300002075.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300013170.mm.1 |
NM_001159513.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013170.mm.1 |
NM_001159513.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013170.mm.1 |
NM_001159514.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013170.mm.1 |
NM_001159514.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013170.mm.1 |
NM_001159515.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013170.mm.1 |
NM_001159515.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013170.mm.1 |
NM_012015.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013170.mm.1 |
NM_012015.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013170.mm.1 |
TC1300002075.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300013167.mm.1 |
NM_001159513.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013167.mm.1 |
NM_001159513.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013167.mm.1 |
NM_001159514.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013167.mm.1 |
NM_001159514.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013167.mm.1 |
NM_001159515.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013167.mm.1 |
NM_001159515.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013167.mm.1 |
NM_012015.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013167.mm.1 |
NM_012015.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013167.mm.1 |
TC1300002075.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300013171.mm.1 |
NM_001159513.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013171.mm.1 |
NM_001159513.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013171.mm.1 |
NM_001159514.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013171.mm.1 |
NM_001159514.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013171.mm.1 |
NM_001159515.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013171.mm.1 |
NM_001159515.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013171.mm.1 |
NM_012015.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013171.mm.1 |
NM_012015.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013171.mm.1 |
TC1300002075.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300013166.mm.1 |
NM_001159513.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013166.mm.1 |
NM_001159513.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013166.mm.1 |
NM_001159514.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013166.mm.1 |
NM_001159514.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013166.mm.1 |
NM_001159515.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013166.mm.1 |
NM_001159515.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013166.mm.1 |
NM_012015.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013166.mm.1 |
NM_012015.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013166.mm.1 |
TC1300002075.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300013154.mm.1 |
NM_001159514.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013154.mm.1 |
NM_001159514.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013154.mm.1 |
NM_001159515.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013154.mm.1 |
NM_001159515.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013154.mm.1 |
TC1300002075.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300013164.mm.1 |
NM_001159513.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013164.mm.1 |
NM_001159513.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013164.mm.1 |
NM_001159514.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013164.mm.1 |
NM_001159514.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013164.mm.1 |
NM_001159515.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013164.mm.1 |
NM_001159515.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013164.mm.1 |
NM_012015.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013164.mm.1 |
NM_012015.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013164.mm.1 |
TC1300002075.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300013172.mm.1 |
NM_001159513.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013172.mm.1 |
NM_001159513.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013172.mm.1 |
NM_001159514.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013172.mm.1 |
NM_001159514.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013172.mm.1 |
NM_001159515.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013172.mm.1 |
NM_001159515.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013172.mm.1 |
NM_012015.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013172.mm.1 |
NM_012015.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300013172.mm.1 |
TC1300002075.mm.1 |
MTA TC |