Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01337824_m1
          See other CRB2 GE Assays ›
          Gene Symbol:
          Crb2
          Gene Name:
          crumbs family member 2
          Gene Aliases:
          5930402A21, BC043114
          Chromosome Location:
          Chr.2: 37776225 - 37799103 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01337824_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Crb2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001163566.1 NP_001157038.1 11 - 12 3591 57
          GenBank mRNA AK031068.1 - 2 - 3 310 57
          BC043114.1 - 10 - 11 3279 57
          BC062128.1 - 4 - 5 1449 57

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200000694.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          cell adhesion molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          mesoderm formation
          somitogenesis
          positive regulation of epithelial to mesenchymal transition
          negative regulation of endopeptidase activity
          notochord formation
          positive regulation of BMP signaling pathway
          maintenance of epithelial cell apical/basal polarity
          response to stimulus
          cardiovascular system development
          calcium ion binding
          aspartic-type endopeptidase inhibitor activity
          enzyme binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P300618 AK031068.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P300618 BC043114.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P300618 BC062128.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P300618 NM_001163566.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P300618 AK031068.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P300618 BC043114.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P300618 BC062128.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P300618 NM_001163566.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 363526 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 692000 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 952046 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 538843 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 294680 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 356024 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 921479 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 898989 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 915549 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 69660 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 819734 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 703166 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 68482 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 128678 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 150215 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 170726 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P113602 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P300618 AK031068.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P300618 BC043114.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P300618 BC062128.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P300618 NM_001163566.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P300618 AK031068.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P300618 BC043114.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P300618 BC062128.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P300618 NM_001163566.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P113602 - -
          Whole Mouse Genome G4122A A_51_P300618 AK031068.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P300618 BC043114.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P300618 BC062128.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P300618 NM_001163566.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P300618 AK031068.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P300618 BC043114.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P300618 BC062128.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P300618 NM_001163566.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P113602 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21144055 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21144098 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21144133 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02109622 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21144086 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02166479 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21144048 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21144125 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21144067 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21144062 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21144080 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21144112 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21144131 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21144076 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21144089 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01756531 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21144071 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21144049 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01925830 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21144042 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21144123 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21144110 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21144115 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21144135 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21144117 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21144053 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21144056 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01384753 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01756533 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21144094 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21144095 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01988920 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21144078 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21144068 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21144047 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21144044 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21144051 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21144142 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21144138 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21144083 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21144061 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01756530 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01977947 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21144087 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21144074 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21144091 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01756534 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21144140 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21144057 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21144050 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21144120 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01756526 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10471828 - -
          Affymetrix Mouse Gene 1.0 ST Array 10471825 - -
          Affymetrix Mouse Gene 1.0 ST Array 10471831 - -
          Affymetrix Mouse Gene 1.0 ST Array 10471821 - -
          Affymetrix Mouse Gene 1.0 ST Array 10471829 - -
          Affymetrix Mouse Gene 1.0 ST Array 10471826 - -
          Affymetrix Mouse Gene 1.0 ST Array 10471830 - -
          Affymetrix Mouse Gene 1.0 ST Array 10471823 - -
          Affymetrix Mouse Gene 1.0 ST Array 10471824 - -
          Affymetrix Mouse Gene 1.0 ST Array 10471832 - -
          Affymetrix Mouse Gene 1.0 ST Array 10471822 - -
          Affymetrix Mouse Gene 1.0 ST Array 10471827 - -
          Affymetrix Mouse Gene 1.1 ST Array 10471819 - -
          Affymetrix Mouse Clariom S Array PSR0200006146.mm.1 NM_001163566.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200006146.mm.1 NM_001163566.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200006146.mm.1 TC0200000694.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200006145.mm.1 NM_001163566.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200006145.mm.1 NM_001163566.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200006145.mm.1 TC0200000694.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200006144.mm.1 NM_001163566.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200006144.mm.1 NM_001163566.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200006144.mm.1 TC0200000694.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 566008 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1258618 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0