Search
Search

Rad51| Gene Symbol: Rad51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011234.4 | NP_035364.1 | 8 - 9 | 997 | 67 | |
| GenBank mRNA | AK011242.1 | - | 8 - 9 | 998 | 67 | |
| AK076468.1 | - | 8 - 9 | 945 | 67 | ||
| AK151157.1 | - | 8 - 9 | 976 | 67 | ||
| AK151177.1 | - | 8 - 9 | 976 | 67 | ||
| AK160282.1 | - | 8 - 9 | 989 | 67 | ||
| BC027384.1 | - | 8 - 9 | 962 | 4951015 | 67 | |
| D13473.1 | - | 8 - 9 | 952 | 67 | ||
| D13803.1 | - | 8 - 9 | 965 | 67 | ||
Set Membership: |
|