Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01338227_g1
          See other OSCAR GE Assays ›
          Gene Symbol:
          Oscar
          Gene Name:
          osteoclast associated receptor
          Gene Aliases:
          mOSCAR, mOSCAR-M1, mOSCAR-M2, mOSCAR-M3
          Chromosome Location:
          Chr.7: 3609813 - 3616157 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01338227_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Oscar
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001290377.1 NP_001277306.1 3 - 4 454 72
          NM_175632.3 NP_783440.1 3 - 4 472 72
          GenBank mRNA AF391159.1 - 3 - 4 430 72
          AF391160.1 - 3 - 4 454 72
          AF391161.1 - 3 - 4 448 72
          BC137776.1 - 3 - 4 399 8860795 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700002131.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Oscar

          Panther Classification:

          Molecular Function -

          immunoglobulin receptor superfamily

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24946359 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1977426 AF391161.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1977426 NM_175632.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1977426 AF391159.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 19452 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 45952 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 41075 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 641 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 674874 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P136593 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P117448 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1977426 AF391161.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1977426 NM_175632.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1977426 AF391159.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946339 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946351 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946334 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946357 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946336 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946353 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946332 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946343 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946344 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946347 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946340 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946337 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946358 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946355 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24946350 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P117448 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P136593 - -
          Whole Mouse Genome G4122A A_51_P404478 BC137776.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P404478 AF391161.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P404478 NM_175632.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P404478 AF391159.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108014 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P117448 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P136593 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24946359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24946329 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946314 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946320 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946329 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946348 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946353 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946334 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946336 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946341 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946332 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946327 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946328 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946344 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946315 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946355 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946337 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946351 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946339 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946317 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946338 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946324 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946340 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946323 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946321 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946330 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946343 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946318 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946325 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946322 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24946350 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10559404 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559402 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559403 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559405 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559400 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559401 - -
          Affymetrix HT MG-430 PM Array Plate 1451710_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1425442_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10559399 - -
          Mouse Genome 430 2.0 Array 1425442_PM_at - -
          Mouse Genome 430 2.0 Array 1425442_at - -
          Mouse Genome 430 2.0 Array 1451710_PM_at - -
          Mouse Genome 430 2.0 Array 1451710_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 232790_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 232790_at - -
          Affymetrix Mouse Clariom S Array PSR0700019175.mm.1 TC0700002131.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700019175.mm.1 NM_175632.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700019176.mm.1 TC0700002131.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700019176.mm.1 NM_175632.3 Refseq
          Mouse Expression Set 430 1425442_PM_at - -
          Mouse Expression Set 430 1451710_PM_at - -
          Mouse Expression Set 430 1451710_at - -
          Mouse Expression Set 430 1425442_at - -
          Mouse Genome 430A 2.0 Array 1451710_PM_at - -
          Mouse Genome 430A 2.0 Array 1425442_PM_at - -
          Mouse Genome 430A 2.0 Array 1451710_at - -
          Mouse Genome 430A 2.0 Array 1425442_at - -
          Affymetrix HT Mouse Genome 430A Array 1451710_at - -
          Affymetrix HT Mouse Genome 430A Array 1451710_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1425442_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1425442_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2120441 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002120441 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1215466 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1215466 - -
          mouse-6 v1.1 (Illumina) scl31835.5.1_93-S - -
          Illumina Sentrix mouse-6.v1 bead array 2120441 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl31835.5.1_93-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2120441 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1215466 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1215466 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline