Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01338424_g1
          See other SOHLH1 GE Assays ›
          Gene Symbol:
          Sohlh1
          Gene Name:
          spermatogenesis and oogenesis specific basic helix-loop-helix 1
          Gene Aliases:
          Gm110, Nohlh
          Chromosome Location:
          Chr.2: 25842995 - 25847248 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01338424_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sohlh1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001001714.1 NP_001001714.1 7 - 8 922 96
          GenBank mRNA AY623913.1 - 7 - 8 922 96
          BC139189.1 - 7 - 8 890 8862211 96
          BC139190.1 - 7 - 8 890 8862212 96

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200003169.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sohlh1

          Panther Classification:

          Molecular Function -

          basic helix-loop-helix transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          ovarian follicle development
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transcription from RNA polymerase II promoter
          multicellular organism development
          spermatogenesis
          regulation of gene expression
          cell differentiation
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          oogenesis
          RNA polymerase II regulatory region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          protein dimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1957543 NM_001001714.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1957543 AY623913.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 623216 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 622779 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 83110 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P102128 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1957543 NM_001001714.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1957543 AY623913.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21074524 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21074549 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21074530 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21074583 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21074542 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21074558 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21074531 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21074539 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21074526 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21074555 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21074566 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21074533 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21074527 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21074554 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21074586 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21074534 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21074537 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21074578 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21074563 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21074581 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21074584 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21074529 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21074561 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21074576 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21074552 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21074569 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21074543 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21074550 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21074546 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21074557 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21074574 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21074523 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21074572 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21074570 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21074525 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21074573 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P102128 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P120712 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130230 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P102128 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02270718 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02270696 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02270687 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02270719 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02270728 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02270721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21074584 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074526 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074524 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02270728 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074593 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074498 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074566 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074573 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074561 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074572 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074480 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074525 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02270682 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074538 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074505 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074581 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074501 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074533 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074549 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074479 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01325841 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02270718 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01731689 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074559 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01731690 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074591 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074531 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074500 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074576 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02270721 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02270719 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074507 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074590 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074596 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074523 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02066502 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074570 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074499 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074574 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02270717 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074554 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074482 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074594 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074555 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074493 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074520 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074578 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074557 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02270708 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01731708 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074586 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074519 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074484 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074539 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074487 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01731726 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074569 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074517 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074587 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074492 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074512 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074563 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074527 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074588 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074544 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074530 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074529 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21074509 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01365175 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02270727 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10480871 - -
          Affymetrix Mouse Gene 1.0 ST Array 10480876 - -
          Affymetrix Mouse Gene 1.0 ST Array 10480874 - -
          Affymetrix Mouse Gene 1.0 ST Array 10480872 - -
          Affymetrix Mouse Gene 1.0 ST Array 10480877 - -
          Affymetrix Mouse Gene 1.0 ST Array 10480870 - -
          Affymetrix Mouse Gene 1.0 ST Array 10480873 - -
          Affymetrix Mouse Gene 1.0 ST Array 10480875 - -
          Affymetrix HT MG-430 PM Array Plate 1460015_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10480869 - -
          Mouse Genome 430 2.0 Array 1460015_at - -
          Mouse Genome 430 2.0 Array 1460015_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 227631_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 227631_at - -
          Affymetrix Mouse Clariom S Array PSR0200026311.mm.1 NM_001001714.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200026311.mm.1 NM_001001714.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200026311.mm.1 TC0200003169.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200026312.mm.1 NM_001001714.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200026312.mm.1 NM_001001714.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200026312.mm.1 TC0200003169.mm.1 MTA TC
          Mouse Expression Set 430 1460015_at - -
          Mouse Expression Set 430 1460015_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1460015_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1460015_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 837751 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1242183 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2850324 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2850324 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0