| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.1 ST Array |
10469609 |
- |
- |
| Mouse Genome 430 2.0 Array |
1439579_PM_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1439579_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR0200001572.mm.1 |
NR_033225.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200001572.mm.1 |
NR_033225.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200001572.mm.1 |
TC0200000336.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200001574.mm.1 |
NR_033225.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200001574.mm.1 |
NR_033225.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200001574.mm.1 |
TC0200000336.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200001575.mm.1 |
NR_033225.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200001575.mm.1 |
NR_033225.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200001575.mm.1 |
TC0200000336.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200001578.mm.1 |
NR_033225.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200001578.mm.1 |
NR_033225.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200001578.mm.1 |
TC0200000336.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200001573.mm.1 |
NR_033225.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200001573.mm.1 |
NR_033225.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200001573.mm.1 |
TC0200000336.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200001576.mm.1 |
NR_033225.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200001576.mm.1 |
NR_033225.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200001576.mm.1 |
TC0200000336.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200001577.mm.1 |
NR_033225.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200001577.mm.1 |
NR_033225.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200001577.mm.1 |
TC0200000336.mm.1 |
MTA TC |
| Affymetrix HT Mouse Genome 430B Array |
1439579_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1439579_PM_at |
- |
- |