Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01338985_s1
          See other ZBTB42 GE Assays ›
          Gene Symbol:
          Zbtb42
          Gene Name:
          zinc finger and BTB domain containing 42
          Gene Aliases:
          EG382639, Gm5188, simiRP58
          Chromosome Location:
          Chr.12: 112678604 - 112682747 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01338985_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zbtb42
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001100460.1 NP_001093930.1 2 1761 91

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          muscle organ development
          nucleic acid binding
          DNA binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P100381 BC069952.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P100381 NM_001100460.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P100381 AK079245.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P100381 BC069952.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P100381 NM_001100460.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P100381 AK079245.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1994832 BC051084.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1994832 BC023876.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1994832 BC046387.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1994832 NM_001100460.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 441016 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P156503 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P100381 BC069952.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P100381 NM_001100460.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P100381 AK079245.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P100381 BC069952.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P100381 NM_001100460.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P100381 AK079245.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1994832 BC051084.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1994832 BC023876.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1994832 BC046387.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1994832 NM_001100460.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P156503 - -
          Whole Mouse Genome G4122A A_52_P100381 BC069952.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P100381 NM_001100460.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P100381 AK079245.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P100381 BC069952.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P100381 NM_001100460.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P100381 AK079245.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P156503 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10398883 - -
          Affymetrix Mouse Gene 1.0 ST Array 10398884 - -
          Affymetrix HT MG-430 PM Array Plate 1456846_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10398881 - -
          Mouse Genome 430 2.0 Array 1456846_at - -
          Mouse Genome 430 2.0 Array 1456846_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 382639_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 382639_at - -
          Affymetrix Mouse Clariom S Array PSR1200009100.mm.1 NM_001100460.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200009100.mm.1 NM_001100460.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200009100.mm.1 TC1200001265.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200009095.mm.1 NM_001100460.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200009095.mm.1 NM_001100460.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200009095.mm.1 TC1200001265.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200009098.mm.1 NM_001100460.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200009098.mm.1 NM_001100460.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200009098.mm.1 TC1200001265.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200009103.mm.1 NM_001100460.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200009103.mm.1 NM_001100460.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200009103.mm.1 TC1200001265.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200009096.mm.1 NM_001100460.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200009096.mm.1 NM_001100460.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200009096.mm.1 TC1200001265.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200009097.mm.1 NM_001100460.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200009097.mm.1 NM_001100460.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200009097.mm.1 TC1200001265.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200009099.mm.1 NM_001100460.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200009099.mm.1 NM_001100460.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200009099.mm.1 TC1200001265.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200009101.mm.1 NM_001100460.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200009101.mm.1 NM_001100460.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200009101.mm.1 TC1200001265.mm.1 MTA TC
          Mouse Expression Set 430 1456846_PM_at - -
          Mouse Expression Set 430 1456846_at - -
          Murine Genome U74v2 Set 129018_at - -
          Murine Genome U74v2 Set 129018_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1456846_at - -
          Affymetrix HT Mouse Genome 430B Array 1456846_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1250745 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0