Search
Search

Olfr1348| Gene Symbol: Olfr1348 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146913.1 | NP_667124.1 | 1 | 269 | 63 | |
| GenBank mRNA | BC120570.1 | - | 1 | 365 | 40129493 | 63 |
| BC137836.1 | - | 1 | 365 | 8860855 | 63 | |
Set Membership: |
|