Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01339377_m1
          See other RGAG1 GE Assays ›
          Gene Symbol:
          Rgag1
          Gene Name:
          retrotransposon gag domain containing 1
          Gene Aliases:
          Gm385, Mar9, Mart9
          Chromosome Location:
          Chr.X: 143057943 - 143105330 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01339377_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rgag1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001040434.2 NP_001035524.2 1 - 2 3988 74
          GenBank mRNA AK220382.1 - 1 - 2 3358 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00001411.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Recommended Assay Mus musculus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          viral or transposable element protein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1983698 AK220382.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1983698 NM_001040434.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 704053 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 774252 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 353596 AK047115.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 378941 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1983698 AK220382.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1983698 NM_001040434.1 mouse_transcriptome
          Agilent Six_ChIP_03_of_03_v2 A_68_P15223190 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14861731 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711690 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15205050 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711660 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711679 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711673 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711644 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711659 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711575 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711603 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711591 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711600 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14812514 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711605 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711684 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711625 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711654 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711663 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711631 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711680 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711627 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711640 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15205093 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711602 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711642 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711685 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711595 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711608 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711717 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711624 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14982893 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14955295 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711657 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711592 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15223183 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711611 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711634 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15205047 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711672 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711584 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711615 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711671 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711619 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711588 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711675 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711593 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711689 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15205036 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711632 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711578 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711648 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15205066 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711617 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711636 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15169874 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15223186 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711621 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14663964 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14688059 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711682 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711653 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711681 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14688062 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711639 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711715 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15166295 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711587 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711650 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14817728 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711716 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711697 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711606 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14955299 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711598 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711694 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711668 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711651 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711665 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711623 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15205100 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711626 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711713 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711687 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15205072 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711628 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711601 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32711696 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10602197 - -
          Affymetrix Mouse Gene 1.1 ST Array 10602196 - -
          Affymetrix Mouse Clariom S Array PSR0X00008936.mm.1 NM_001040434.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00008936.mm.1 TC0X00001411.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00008935.mm.1 NM_001040434.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00008935.mm.1 TC0X00001411.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00008933.mm.1 NM_001040434.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00008933.mm.1 TC0X00001411.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00008934.mm.1 NM_001040434.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00008934.mm.1 TC0X00001411.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 525518 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline