Search
Search

Adck1| Gene Symbol: Adck1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277296.1 | NP_001264225.1 | 9 - 10 | 1486 | 63 | |
| NM_001277297.1 | NP_001264226.1 | 9 - 10 | 1298 | 63 | ||
| NM_028105.4 | NP_082381.1 | 9 - 10 | 1407 | 63 | ||
| GenBank mRNA | AK011310.1 | - | 9 - 10 | 1258 | 63 | |
| AK050868.1 | - | 9 - 10 | 1388 | 63 | ||
| AK145987.1 | - | 6 - 7 | 966 | 63 | ||
| AK154839.1 | - | 9 - 10 | 1353 | 63 | ||
| AK218268.1 | - | 1 - 2 | 56 | 63 | ||
| BC010539.1 | - | 9 - 10 | 1300 | 4162365 | 63 | |
Set Membership: |
|