Search
Search

Topaz1| Gene Symbol: Topaz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199736.1 | NP_001186665.1 | 18 - 19 | 4558 | 63 | |
| GenBank mRNA | HM631980.1 | - | 18 - 19 | 4558 | 63 | |
Set Membership: |
|