Search
Search

Ttc37| Gene Symbol: Ttc37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081352.1 | NP_001074821.1 | 21 - 22 | 2499 | 60 | |
| GenBank mRNA | AK162476.1 | - | 1 - 2 | 690 | 60 | |
| BC035338.1 | - | 2 - 3 | 788 | 60 | ||
| EF591876.1 | - | 2003 | 60 | |||
Set Membership: |
|