Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01341415_m1
          See other CFP GE Assays ›
          Gene Symbol:
          Cfp
          Gene Name:
          complement factor properdin
          Gene Aliases:
          BCFG, Pfc
          Chromosome Location:
          Chr.X: 20925535 - 20931525 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01341415_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cfp
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008823.3 NP_032849.2 3 - 4 462 70
          GenBank mRNA AK150212.1 - 3 - 4 423 70
          AK152782.1 - 3 - 4 413 70
          AK154306.1 - 3 - 4 446 70
          AK159291.1 - 3 - 4 463 70
          AK159609.1 - 3 - 4 413 70
          AK159853.1 - 3 - 4 413 70
          AK171374.1 - 3 - 4 446 70
          BC132369.1 - 3 - 4 446 40130646 70
          BC138306.1 - 3 - 4 446 8861326 70
          X12905.1 - 2 - 3 311 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00001934.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cfp

          Gene Ontology Categories:

          Function(s)

          immune system process
          complement activation, alternative pathway
          innate immune response

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P408363 AK171374.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P408363 BC132369.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P408363 BC138306.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P408363 X12905.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P408363 AK154306.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P408363 AK171374.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P408363 BC132369.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P408363 BC138306.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P408363 X12905.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P408363 AK154306.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 299074 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 822345 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 629377 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 340468 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P172463 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P113244 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P408363 AK171374.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P408363 BC132369.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P408363 BC138306.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P408363 X12905.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P408363 AK154306.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P408363 AK171374.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P408363 BC132369.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P408363 BC138306.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P408363 X12905.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P408363 AK154306.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32290709 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32290698 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32290723 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32290722 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32290662 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32290694 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32290714 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32290705 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32290718 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32290689 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32290711 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32290692 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32290715 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32290702 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32290696 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32290695 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32290713 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32290701 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32290724 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32290690 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32290664 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32290716 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32290699 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32290721 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32290666 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32290720 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32290707 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32290710 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P172463 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P113244 - -
          Whole Mouse Genome G4122A A_51_P408363 AK171374.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P408363 BC132369.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P408363 BC138306.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P408363 X12905.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P408363 AK154306.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P408363 AK171374.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P408363 BC132369.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P408363 BC138306.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P408363 X12905.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P408363 AK154306.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P113244 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P172463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32290607 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14613283 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32290698 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32290684 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32290673 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32290685 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14703534 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14613286 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15041497 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32290711 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32290694 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15041512 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15041500 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32290713 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32290680 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32290686 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15071120 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32290670 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14703538 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32290699 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14967478 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32290665 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32290695 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32290676 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32290690 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32290702 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32290696 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32290668 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14703532 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32290714 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32290664 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32290675 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10603861 - -
          Affymetrix Mouse Gene 1.0 ST Array 10603867 - -
          Affymetrix Mouse Gene 1.0 ST Array 10603865 - -
          Affymetrix Mouse Gene 1.0 ST Array 10603862 - -
          Affymetrix Mouse Gene 1.0 ST Array 10603868 - -
          Affymetrix Mouse Gene 1.0 ST Array 10603866 - -
          Affymetrix Mouse Gene 1.0 ST Array 10603864 - -
          Affymetrix Mouse Gene 1.0 ST Array 10603863 - -
          Affymetrix Mouse Gene 1.0 ST Array 10603869 - -
          Affymetrix HT MG-430 PM Array Plate 1452279_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10603860 - -
          Mouse Genome 430 2.0 Array 1452279_PM_at - -
          Mouse Genome 430 2.0 Array 1452279_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 18636_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 18636_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0X00012271.mm.1 NM_008823.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00012271.mm.1 TC0X00001934.mm.1 MTA TC
          Mouse Expression Set 430 1452279_at - -
          Mouse Expression Set 430 1452279_PM_at - -
          Murine Genome U74v2 Set 101468_at - -
          Murine Genome U74v2 Set 101468_PM_at - -
          Mouse Genome 430A 2.0 Array 1452279_at - -
          Mouse Genome 430A 2.0 Array 1452279_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1452279_at - -
          Affymetrix HT Mouse Genome 430A Array 1452279_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 883690 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1228320 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1228320 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0