Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01341635_m1
          See other CORO2A GE Assays ›
          Gene Symbol:
          Coro2a
          Gene Name:
          coronin, actin binding protein 2A
          Gene Aliases:
          9030208C03Rik, AI563590, IR10
          Chromosome Location:
          Chr.4: 46536937 - 46601929 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01341635_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Coro2a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001164804.1 NP_001158276.1 2 - 3 319 91
          NM_178893.4 NP_849224.2 3 - 4 544 91
          GenBank mRNA AK030097.1 - 3 - 4 547 91
          AK044060.1 - 2 - 3 281 91
          AK054394.1 - 2 - 3 320 91
          AK152490.1 - 2 - 3 308 91
          AK153134.1 - 2 - 3 308 91
          AK154247.1 - 2 - 3 276 91
          AK170184.1 - 2 - 3 277 91
          BC138731.1 - 2 - 3 344 8861753 91
          BC138732.1 - 2 - 3 344 8861754 91

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400002603.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Coro2a

          Panther Classification:

          Molecular Function -

          non-motor actin binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          actin cytoskeleton organization
          actin binding
          actin filament binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0400021249.mm.1 TC0400002603.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400021249.mm.1 NM_001164804.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400021236.mm.1 TC0400002603.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400021236.mm.1 NM_001164804.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400021236.mm.1 NM_001164804.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400021236.mm.1 NM_178893.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400021236.mm.1 NM_178893.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0400021237.mm.1 TC0400002603.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400021237.mm.1 NM_001164804.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400021237.mm.1 NM_001164804.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400021237.mm.1 NM_178893.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400021237.mm.1 NM_178893.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0400021241.mm.1 TC0400002603.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400021241.mm.1 NM_178893.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400021241.mm.1 NM_178893.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0400021253.mm.1 TC0400002603.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400021253.mm.1 NM_001164804.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400021239.mm.1 TC0400002603.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400021239.mm.1 NM_178893.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400021239.mm.1 NM_178893.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0400021240.mm.1 TC0400002603.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400021240.mm.1 NM_178893.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400021240.mm.1 NM_178893.4 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 534213 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline