Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01341836_m1
          See other AVL9 GE Assays ›
          Gene Symbol:
          Avl9
          Gene Name:
          AVL9 homolog (S. cerevisiase)
          Gene Aliases:
          5830411G16Rik, D730049P16Rik, mKIAA0241
          Chromosome Location:
          Chr.6: 56714905 - 56761912 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01341836_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Avl9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_030235.1 NP_084511.1 15 - 16 2052 100
          GenBank mRNA AK020003.1 - 15 - 16 2029 100
          AK083605.1 - 15 - 16 2009 100
          AK122228.1 - 15 - 16 2030 100
          AK147365.1 - 15 - 16 2054 100
          BC117878.1 - 15 - 16 2014 40095961 100
          BC117879.1 - 15 - 16 2013 100

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600000676.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          cell migration
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24476448 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24476452 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24476446 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24476451 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24476445 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24476447 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P480402 AK160353.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P480402 AK122228.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P480402 NM_030235.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P480402 AK014039.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P480402 AK160353.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P480402 AK122228.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P480402 NM_030235.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P480402 AK014039.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 623236 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 77292 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 105223 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 701276 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 473397 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 694518 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 413910 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 262143 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 772647 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 175997 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 57395 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 390205 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 345868 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 432278 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 255059 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 707850 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 410854 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 556183 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 424665 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 805933 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 960130 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 299872 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 323999 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 568563 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 413907 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 165807 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 122372 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 27516 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 96435 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 15759 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 64656 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P111934 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P157224 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P480402 AK160353.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P480402 AK122228.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P480402 NM_030235.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P480402 AK014039.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P480402 AK160353.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P480402 AK122228.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P480402 NM_030235.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P480402 AK014039.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05910305 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05910285 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P157224 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P111934 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05910366 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01337327 - -
          Whole Mouse Genome G4122A A_51_P220852 BC117878.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P220852 BC117879.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P220852 AK020003.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P220852 AK083605.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P220852 AK122228.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P480402 AK160353.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P480402 AK122228.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P480402 NM_030235.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P480402 AK014039.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P480402 AK160353.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P480402 AK122228.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P480402 NM_030235.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P480402 AK014039.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P157224 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P01337327 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P111934 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24476447 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24476452 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24476445 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24476451 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24476448 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24476446 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476653 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05062764 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476459 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476480 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476469 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476646 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476650 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476477 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476651 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476468 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476449 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05732602 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05078323 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476645 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476644 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476473 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476460 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476642 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476640 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476648 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476462 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476641 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476647 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24476463 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10538541 - -
          Affymetrix Mouse Gene 1.0 ST Array 10538529 - -
          Affymetrix Mouse Gene 1.0 ST Array 10538534 - -
          Affymetrix Mouse Gene 1.0 ST Array 10538528 - -
          Affymetrix Mouse Gene 1.0 ST Array 10538530 - -
          Affymetrix Mouse Gene 1.0 ST Array 10538532 - -
          Affymetrix Mouse Gene 1.0 ST Array 10538543 - -
          Affymetrix Mouse Gene 1.0 ST Array 10538527 - -
          Affymetrix Mouse Gene 1.0 ST Array 10538542 - -
          Affymetrix Mouse Gene 1.0 ST Array 10538540 - -
          Affymetrix Mouse Gene 1.0 ST Array 10538531 - -
          Affymetrix Mouse Gene 1.0 ST Array 10538535 - -
          Affymetrix Mouse Gene 1.0 ST Array 10538536 - -
          Affymetrix Mouse Gene 1.0 ST Array 10538539 - -
          Affymetrix Mouse Gene 1.0 ST Array 10538533 - -
          Affymetrix Mouse Gene 1.0 ST Array 10538537 - -
          Affymetrix Mouse Gene 1.0 ST Array 10538538 - -
          Affymetrix HT MG-430 PM Array Plate 1433177_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1434659_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1430144_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1457023_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10538526 - -
          Mouse Genome 430 2.0 Array 1433177_at - -
          Mouse Genome 430 2.0 Array 1457023_PM_at - -
          Mouse Genome 430 2.0 Array 1430144_PM_at - -
          Mouse Genome 430 2.0 Array 1433177_PM_at - -
          Mouse Genome 430 2.0 Array 1457023_at - -
          Mouse Genome 430 2.0 Array 1434659_at - -
          Mouse Genome 430 2.0 Array 1434659_PM_at - -
          Mouse Genome 430 2.0 Array 1430144_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 78937_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 78937_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0600005249.mm.1 TC0600000676.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600005249.mm.1 NM_030235.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600005249.mm.1 NM_030235.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600005248.mm.1 TC0600000676.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600005248.mm.1 NM_030235.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600005248.mm.1 NM_030235.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600005251.mm.1 TC0600000676.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600005251.mm.1 NM_030235.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600005251.mm.1 NM_030235.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600005250.mm.1 TC0600000676.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600005250.mm.1 NM_030235.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600005250.mm.1 NM_030235.1 Refseq
          Mouse Expression Set 430 1434659_PM_at - -
          Mouse Expression Set 430 1434659_at - -
          Mouse Expression Set 430 1457023_PM_at - -
          Mouse Expression Set 430 1430144_PM_at - -
          Mouse Expression Set 430 1430144_at - -
          Mouse Expression Set 430 1457023_at - -
          Mouse Genome 430A 2.0 Array 1434659_at - -
          Mouse Genome 430A 2.0 Array 1434659_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1434659_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1434659_at - -
          Affymetrix HT Mouse Genome 430B Array 1430144_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1457023_at - -
          Affymetrix HT Mouse Genome 430B Array 1433177_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1457023_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1433177_at - -
          Affymetrix HT Mouse Genome 430B Array 1430144_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_2709585 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1233082 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline